missing translation for 'onlineSavingsMsg'
Learn More

Honeywellâ„¢ Millerâ„¢ H500 Safety Harness Series

Synthesizes safety, quality and comfort. The harness is designed around ergonomic safety, usability, style and performance, aiming to keep workers safe, supported and make their jobs easier.

$209.99 - $230.66

Specifications

Certifications/Compliance OSHA 1926.502, ANSI Z359.11, CSA Z259.10
Color Green
Product Type Full body harness
For Use With (Equipment) Anchorage connector, Self-retracting lifeline, Lanyard
View More Specs

Products 4
Catalog Number Mfr. No. Buckle Style (Chest) Buckle Style (Leg) Material Size Price Quantity  
Catalog Number Mfr. No. Buckle Style (Chest) Buckle Style (Leg) Material Size Price Quantity  
19-080-005
View Documents
Honeywellâ„¢
H5CC222022
Quick-connect chest buckles Quick-connect leg buckles Polyester, Nylon, Aluminum Universal
Each for $230.66
Only null left
 
19-080-017
View Documents
Honeywellâ„¢
H5CC311122
Mating chest buckles Tongue leg buckles Polyester, Nylon, Steel Universal
Each for $209.99
Only null left
 
19-080-016
View Documents
Honeywellâ„¢
H5CC311121
Mating chest buckles Tongue leg buckles Polyester, Nylon, Steel Small/Medium
Each for $209.99
Only null left
 
19-080-018
View Documents
Honeywellâ„¢
H5CC311123
Mating chest buckles Tongue leg buckles Polyester, Nylon, Steel 2X-Large
Each for $209.99
Only null left
 
Description

Description

  • Specially formulated webbing flexes to accomodate movement and body contours for optimum comfort around legs and shoulders
  • Highly breathable, lightweight, ergonomic shoulder/back padding minimizes overhead muscular fatigue
    • Keeps workers drier and cooler
  • Robust and ergonomic pressure-relief waist pad with adequate foam density for optimum lumbar support
  • Smart anti-tangle for hassle-free donning experience
  • User friendly resettable lanyard keepers
  • Clear identifiable smart impact indicators for easier inspection
  • Rated for up to a 420 lb. weight capacity
Specifications

Specifications

OSHA 1926.502, ANSI Z359.11, CSA Z259.10
Green
Full body harness
Anchorage connector, Self-retracting lifeline, Lanyard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'