Learn More
Honeywell™ Miller™ QuickPick™ Rescue Kit
Rescue Kits deliver peace of mind during unexpected, stressful peer-rescue situations
Supplier:  Honeywell™ QP25FT
 View more versions of this product
Catalog No. 19-826-900
Only null left
Explore available promotions
Description
Description
The rescuer can remotely attach the system to the suspended worker while remaining securely anchored on the walking/working surface. All rescue components are contained in one easy-to-store kit with no assembly necessary — only requires an overhead anchorage.
  • Out-of-the-bag solution for prompt rescue, with no assembly required
  • Quickly raise or lower a fallen worker to safety — avoid suspension trauma
  • Professional rescue solution; enhances worker safety and corporate rescue plan
Applications:
Construction, Manufacturing, Municipal Services, Oil and Gas, Transportation, Utilities, Wind Energy
Compliance
Meets ANSI Z359.4 requirements.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
    • Certifications and Seals of Approval
Specifications
Specifications
10.3kg
QuickPick Premium Kit
181.4 kg
400 lb.
25 ft.
22.9 lb.
OSHA, ANSI A10.32, ANSI Z359.4
7.62 m
Product Suggestions
Product Suggestions