missing translation for 'onlineSavingsMsg'
Learn More

Millennium Sciences™ Cell-VU Prestained Slides

Catalog No. DRM900
Change view
Click to view available options
Stain Type:
Methylene Blue and Cresyl Violet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stain Type
DRM900 Methylene Blue and Cresyl Violet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DRM900 Supplier Millennium Sciences™ Supplier No. DRM900

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for simplified and convenient manual hematology staining that is safer with less exposure to reagents and stains

  • CELL-VU™ pre-stained Hematology/Morphology slides combine conventional microscope slides with a combination of methylene blue and cresyl violet stains into a single, ready to use device.
  • Samples are applied directly to the slide and a coverslip is placed over the sample. One step process limits exposure to reagents and potentially harmful stains.
  • Each slide contains a dried layer of stain that allows for rapid visualization and differential cell assessment from whole blood and body fluids such as CSF, and urine samples.
  • CELL-VU™ can also be used for World Health Organization and Kruger strict morphology sperm assessment from undiluted semen.

Specifications

Length (Metric) 76.2 mm
Material Glass
Width (Metric) 25.4 mm
Type Pre-Stained Morphology Slides
Stain Type Methylene Blue and Cresyl Violet
Regulatory Status FDA registered
Color Blue
Length (English) 3 in.
Quantity 75
Width (English) 1 in.