Learn More
Sarstedt Inc MICRO TUBES 0.5ML TYPE A 250PK DFS Item
Supplier:  Sarstedt Inc 72730217
Micro tube 0.5 ml, PP, with knurls, with skirted base, with writing space, with attached unscrewed screw cap, Biosphere� plus certified (sterile, DNA, RNase, and ATP-free and non-pyrogenic), 25 pieces packed per bag
Catalog No. NC0443119
Only null left
Explore available promotions
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Product Suggestions
Product Suggestions