Learn More
Methyl 4-methoxycarbonylbenzoylacetate, 95%, Thermo Scientificâ„¢
$58.13 - $58.13
Chemical Identifiers
CAS | 22027-52-7 |
---|---|
Molecular Formula | C13H14O5 |
Molecular Weight (g/mol) | 250.25 |
MDL Number | MFCD00800637 |
InChI Key | GFDCYWYWNSTMDX-UHFFFAOYSA-N |
PubChem CID | 3864207 |
IUPAC Name | methyl 4-(3-methoxy-3-oxopropanoyl)benzoate |
SMILES | CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC325710010
|
Thermo Scientific Chemicals
325710010 |
1 g | Glass bottle |
Each for $58.13
|
|
|||||
Chemical Identifiers
22027-52-7 | |
250.25 | |
GFDCYWYWNSTMDX-UHFFFAOYSA-N | |
methyl 4-(3-methoxy-3-oxopropanoyl)benzoate |
C13H14O5 | |
MFCD00800637 | |
3864207 | |
CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC |
Specifications
22027-52-7 | |
White to Yellow | |
94% min | |
C13H14O5 | |
1 g | |
Solubility in water: insoluble in water. Other solubilities: slightly soluble in acetone,methanol | |
CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC | |
250.25 | |
236.22 | |
Powder |
81°C to 86°C | |
Authentic | |
Glass bottle | |
MFCD00800637 | |
methyl 4-3-methoxy-3-oxopropanoyl benzoate, methyl 4-methoxycarbonylbenzoylacetate, 4-2-methoxycarbonyl-acetyl-benzoic acid methyl ester, 4-methoxycarbonyl-beta-oxobenzenepropanoic acid methyl ester, benzenepropanoic acid,4-methoxycarbonyl-a-oxo-, methyl ester, acmc-20efie, methyl 4-methoxycarbonyl benzoyl acetate, methyl4-methoxycarbonylbenzoylacetate, 4-methoxycarbonylbenzoyl acetic acid methyl ester, methyl 3-4-methoxycarbonyl phenyl-3-oxopropanoate | |
GFDCYWYWNSTMDX-UHFFFAOYSA-N | |
methyl 4-(3-methoxy-3-oxopropanoyl)benzoate | |
3864207 | |
95% | |
Methyl 4-methoxycarbonylbenzoylacetate |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only