missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange, Sodium Salt, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 269404
Description
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Methyl Orange, Sodium Salt | |
C14H14N3NaO3S | |
STZCRXQWRGQSJD-UHFFFAOYSA-M | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate | |
23673835 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 1.00kg | |
Powder |
547-58-0 | |
C.I. 13025 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] | |
327.334 | |
327.34 | |
Wide Mouth Amber Glass Bottle | |
125 g |