missing translation for 'onlineSavingsMsg'
Learn More

Mesitylene, 99%, Extra Pure

CAS: 108-67-8 | C9H12 | 120.19 g/mol

Supplier:  Thermo Scientific Chemicals 125580025

 View more versions of this product

Catalog No. AC125580025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.<br

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

GHS Signal Word: Danger

EINECSNumber : 203-604-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only