Learn More
Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)
CAS: 15364-94-0 | Cl2H12MnO14 | 361.92 g/mol
Supplier: Thermo Scientific Chemicals 04431818
Description
Manganese(II) perchlorate hexahydrate, is a good oxidizing agent and also acts as an intermediates in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Manganese (II) perchlorate hexahydrate | |
15364-94-0 | |
50 g | |
Cl2H12MnO14 | |
UN1481 | |
Soluble in water,benzene,ethyl acetate | |
O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
361.92 | |
361.93 (253.84 Anhydrous) | |
Odorless |
165°C (decomposition) | |
Crystalline | |
(metals basis) | |
MFCD00150255 | |
manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent grade | |
AIFGVOMWPFMOCN-UHFFFAOYSA-L | |
manganese(2+);diperchlorate;hexahydrate | |
203855 | |
99.995% | |
Hygroscopic |
Chemical Identifiers
15364-94-0 | |
361.92 | |
AIFGVOMWPFMOCN-UHFFFAOYSA-L | |
203855 | |
O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Cl2H12MnO14 | |
MFCD00150255 | |
manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent gradeShow More | |
manganese(2+);diperchlorate;hexahydrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P280-P370+P378q-P501c
H272
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber : 237-390-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only