missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) perchlorate hexahydrate, 99.995% (metals basis)

CAS: 15364-94-0 | Cl2H12MnO14 | 361.92 g/mol

Supplier:  Thermo Scientific Chemicals 04431809

 View more versions of this product

Catalog No. AA4431809


Only null left
Explore available promotions

Description

Description

Manganese(II) perchlorate hexahydrate, is a good oxidizing agent and also acts as an intermediates in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P280-P370+P378q-P501c

H272

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

EINECSNumber : 237-390-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only