missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium citrate, dibasic hydrate
CAS: 119851-23-9 | C6H6MgO7 | 214.41 g/mol
$199.48 - $568.42
Chemical Identifiers
| CAS | 119851-23-9 |
|---|---|
| Molecular Formula | C6H6MgO7 |
| Molecular Weight (g/mol) | 214.41 |
| MDL Number | MFCD00151706 |
| InChI Key | DIXGJWCZQHXZNR-UHFFFAOYNA-L |
| Synonym | magnesium 2-carboxymethyl-2-hydroxysuccinate hydrate |
| PubChem CID | 88316079 |
| SMILES | [Mg++].OC(=O)CC(O)(CC([O-])=O)C([O-])=O |
Description
Magnesium citrate is used to clean stool from the intestines before surgery. It is also used as a laxative for relief of constipation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 119851-23-9 | |
| 214.41 | |
| DIXGJWCZQHXZNR-UHFFFAOYNA-L | |
| 88316079 |
| C6H6MgO7 | |
| MFCD00151706 | |
| magnesium 2-carboxymethyl-2-hydroxysuccinate hydrate | |
| [Mg++].OC(=O)CC(O)(CC([O-])=O)C([O-])=O |
Specifications
| 119851-23-9 | |
| 250 g | |
| MFCD00151706 | |
| magnesium 2-carboxymethyl-2-hydroxysuccinate hydrate | |
| DIXGJWCZQHXZNR-UHFFFAOYNA-L | |
| magnesium(2+) 2-(carboxymethyl)-2-hydroxybutanedioate | |
| 88316079 | |
| Magnesium citrate, dibasic hydrate |
| Powder | |
| C6H6MgO7 | |
| 14,5663 | |
| Slightly soluble in water. | |
| [Mg++].OC(=O)CC(O)(CC([O-])=O)C([O-])=O | |
| 214.41 | |
| 214.14 (Anhydrous) |
Safety and Handling
EINECSNumber : 231-923-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only