Learn More
Magnesium Aluminum Silicate, Type IA, NF, Spectrum™ Chemical
M1021, 1327-43-1
Supplier:  Spectrum Chemical Mfg Cor M102112KG
 View more versions of this product
Catalog No. 18604013
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Magnesium Aluminum Silicate, Type IA, NF is used in the pharmaceutical formulation process as an absorbent, anticaking agent, opacifying agent, slip modifier, and an aqueous viscosity increasing agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
1327-43-1
9.0-10.0
NF
Fiber Drum
100%
12 kg
8%
Chemical Identifiers
1327-43-1
Product Suggestions
Product Suggestions