missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Machine and Equipment Label, Legend: LOCK-OUT HERE
Communicates important safety information and machine operational instructions. Brady Machine and Equipment Labels are made of durable, self-sticking polyester and most feature pictograms as well as text to inform safety procedures and potential hazards with machine operation.
Supplier: Brady™ 86245
Description
- Placed on machine or equipment to increase safety
- Made of B-302 self sticking polyester
- Available in size (W x H): 12.7 x 8.89cm (5 x 3.5 in.)
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Acrylic, Permanent, Cold-temperature | |
Gloss | |
Machine/Equipment Label | |
English | |
Rectangle | |
127 x 88.9 mm (W x H) |
Black, Red, White | |
Self-adhesive Polyester (B-302) | |
Legend: LOCK-OUT HERE | |
5/Pk. | |
5 x 3.5 in. (W x H) | |
0.01 in. |