Learn More
Lithium perchlorate, anhydrous, 99%
For Lithium Battery | CAS: 7791-03-9 | ClH6LiO7 | 160.43 g/mol
Supplier: Thermo Scientific Chemicals 04422530
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
400°C (decomposition) | |
236°C | |
White | |
250 g | |
ClH6LiO7 | |
lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1 | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
lithium;perchlorate | |
23665649 | |
Odorless | |
White crystals |
Lithium perchlorate, anhydrous | |
7791-03-9 | |
Granules | |
99% | |
MFCD00011079 | |
Soluble in water,alcohol and ethyl acetate. | |
[Li+].[O-]Cl(=O)(=O)=O | |
160.43 | |
106.4 | |
Hygroscopic | |
2.43 |
Chemical Identifiers
7791-03-9 | |
160.43 | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
23665649 | |
[Li+].[O-]Cl(=O)(=O)=O |
ClH6LiO7 | |
MFCD00011079 | |
lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1Show More | |
lithium;perchlorate |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P283-P302+P352-P304+P340-P305+P351+P338-P306+P360-P312-P332+P313-P363-P370+P378q-P371+P380+P375-P501c
H271-H315-H319-H335
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 232-237-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only