Learn More
Lithium metaborate, anhydrous, 99.9% (metals basis)
CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol
Supplier: Thermo Scientific Chemicals 01259118
Description
Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium metaborate, anhydrous | |
13453-69-5 | |
50 g | |
B4H14Li2O7 | |
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs | |
DPYKRXYVXDYLEY-UHFFFAOYSA-N | |
tetraboron heptahydrate dilithium | |
123308 | |
Hygroscopic |
845°C | |
Powder | |
99.9% (metals basis) | |
MFCD00011089 | |
Soluble in ethanol. Partially soluble in cold water. | |
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O | |
183.23 | |
49.75 | |
1.397 |
Chemical Identifiers
13453-69-5 | |
183.23 | |
DPYKRXYVXDYLEY-UHFFFAOYSA-N | |
123308 |
B4H14Li2O7 | |
MFCD00011089 | |
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs | |
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O |
Safety and Handling
EINECSNumber : 236-631-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only