missing translation for 'onlineSavingsMsg'
Learn More

Lithium dimethylamide, 96%

CAS: 3585-33-9 | C2H6LiN | 51.017 g/mol

Supplier:  Thermo Scientific Chemicals 04414514

 View more versions of this product

Catalog No. AA4414514


Only null left
Explore available promotions

Description

Description

It is used in the synthesis of some d0 organoimido complexes of the early transition metals and in meta rearrangement in the reaction of triphenyl-(p-bromophenyl)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H250-H261-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P222-P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H250-H261-H314-H335

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC, WATER-REACTIVE

EINECSNumber : 222-714-3

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only