missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium bis(trimethylsilyl)amide, 1.0M sol. in methyl tert-butyl ether, AcroSeal™

CAS: 4039-32-1 | C6H18LiNSi2 | 167.33 g/mol
$336.45 - $336.45
Specifications
Chemical Name or Material | Lithium bis(trimethylsilyl)amide |
---|---|
Name Note | 1.0M Solution in Methyl tert-Butyl Ether |
CAS | 4039-32-1,1634-04-4 |
Color | Brown to Yellow |
Density | 0.8000g/mL |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC345671000
|
Thermo Scientific Chemicals
345671000 |
100 mL | AcroSeal™ Glass bottle |
Each for $336.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium bis(trimethylsilyl)amide | |
4039-32-1,1634-04-4 | |
0.8000g/mL | |
−28°C | |
MFCD00008261 | |
0.8 | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
lithium;bis(trimethylsilyl)azanide | |
2733832 | |
21 to 25% active base (as LiNSi) |
1.0M Solution in Methyl tert-Butyl Ether | |
Brown to Yellow | |
55.0°C to 56.0°C | |
C6H18LiNSi2 | |
04,296; 05,393; 07,197; 12,280; 13,165; 14,194 | |
lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C | |
167.33 | |
167.33 | |
Solution |
RUO – Research Use Only