Learn More
LiChropur™ Silylating Mixture I, According to Sweeley, For GC Derivatization, MilliporeSigma™ Supelco™
Useful silylating mixture for the silylation of hydroxy-compounds
Supplier:  Merck Emd Millipore 8543110ML
 View more versions of this product
Catalog No. 111019014
Only null left
Explore available promotions
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
318974-69-5
10 mL
For GC derivatization
MFCD00213951
Chlorotrimethylsilane - hexamethyldisilazane mixture; Sylon(TM) HTP; hexamethyldisilazane/chlorotrimethylsilane/pyridine = 2:1:10 (v/v/v); HMDS + TMCS
Sylon
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany