Learn More
Lead(II) perchlorate trihydrate, 99%
CAS: 13453-62-8 | Cl2H6O11Pb | 460.10 g/mol
Supplier: Thermo Scientific Chemicals 254592500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent- Environmental benefits include:
- Renewable Energy
Specifications
Lead(II) perchlorate trihydrate | |
99% | |
White to Beige | |
Adhering Crystals | |
98.5% min. (Complexometry) | |
Pb(ClO4)2·3H2O | |
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent | |
KKGGEAQRICYXNM-UHFFFAOYSA-L | |
lead(2+);diperchlorate;trihydrate | |
25021834 | |
99% | |
2.6 |
100.0°C | |
13453-62-8 | |
3 to 5 (5% soln.) | |
250 g | |
Cl2H6O11Pb | |
MFCD00149817 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
460.10 | |
460.14 | |
Glass bottle | |
2.6000g/mL |
Chemical Identifiers
13453-62-8 | |
460.10 | |
KKGGEAQRICYXNM-UHFFFAOYSA-L | |
25021834 |
Cl2H6O11Pb | |
MFCD00149817 | |
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagentShow More | |
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.
May intensify fire; oxidizer.
GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only