missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ LabKlenz 250 Acid Detergent

LabKlenz 250 Acid Detergent is a low-foaming, citric acid-based, liquid detergent. It is formulated for use as a rinse aid, as well as for the removal of particulate, scale and inorganic soils. pH (1%): 2.6, typical.

$654.08 - $758.03

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 2.6
Scent Unscented
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-467-588
View Documents
STERIS™
1L2508
1 gal.
Case of 4 Each for $654.08
Only null left
 
17-467-589
View Documents
STERIS™
1L2505
5 gal.
Each for $758.03
Only null left
 
Description
Show More
Specifications

Specifications

Detergent
Clear Liquid
Unscented
No
2.6
Non-sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'