missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Histidine hydrochloride monohydrate, 98%

CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol
$59.54 - $785.79
Chemical Identifiers
CAS | 5934-29-2 |
---|---|
Molecular Formula | C6H12ClN3O3 |
Molecular Weight (g/mol) | 209.63 |
MDL Number | MFCD00151027 |
InChI Key | CMXXUDSWGMGYLZ-XRIGFGBMSA-N |
Synonym | l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate |
PubChem CID | 165377 |
IUPAC Name | (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride |
SMILES | O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC411730250
|
Thermo Scientific Chemicals
411730250 |
25 g | Plastic Bottle |
Each for $59.54
|
|
|||||
AC411731000
|
Thermo Scientific Chemicals
411731000 |
100 g | Plastic Bottle |
Each for $165.37
|
|
|||||
AC411735000
|
Thermo Scientific Chemicals
411735000 |
500 g | Plastic Bottle |
Each for $785.79
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5934-29-2 | |
209.63 | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
165377 | |
O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O |
C6H12ClN3O3 | |
MFCD00151027 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
(2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride |
Specifications
200ppm max. | |
3 to 5 (2.5% soln. at 25°C) | |
10ppm max. | |
10ppm max. | |
98% | |
C6H12ClN3O3 | |
MFCD00151027 | |
25, 513 | |
10, 4617 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O | |
+ 9.90 (20.00°C c=11,6 N HCl) | |
25 g | |
209.64 | |
Crystalline Powder or Crystals |
5934-29-2 | |
White | |
Authentic | |
7% to 10% (3 hrs) (145 to 150°C) | |
0.5% max. (TLC) | |
Plastic Bottle | |
+ 9.90 | |
300ppm max. | |
0.1% max. | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
(2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride | |
209.63 | |
165377 | |
98% | |
L-Histidine hydrochloride monohydrate, 98% |
Safety and Handling
RTECSNumber : MS3119000
RUO – Research Use Only