missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Koehler™ Instrument Replacement Borosilicate Glass Flash Point Cup for Tag Open Tester
Supplier: Koehler™ Instrument K15520
Description
Greener Choice Claims
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Certifications and Seals of Approval
- Renewable Energy
- Source Reduction
Specifications
Flash Point Test Cup | |
Glass | |
Tag Open Tester |