Learn More
Ansell KleenGuard™ A20 Breathable Particle Protection Lab Coats

A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.
Supplier: Ansell 10019
Description
A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.
- Tailored style made of breathable patented MICROFORCE™ Barrier SMS Fabric
- Knee-length, serged seams
- Long sleeves, notched-lapel collar
- Air and moisture vapor can pass through to aid wearer comfort
- Meets NFPA 99 criteria for antistatic materials
- Two styles to choose from:
- Open wrists with pockets, snap closure
- Elastic wrists without pockets, hook and loop closure
Compliance
NFPA 99 compliant antistatic material
- Environmental benefits include:
- Recyclable
Specifications
General Purpose Lab Coat | |
Unisex | |
White | |
Open | |
Non-sterile | |
2 | |
10019 | |
NFPA 99 | |
37.5 in. | |
1.2 oz. Fabric |
Medium | |
SMS | |
Snaps | |
Limited-use | |
Knee-length | |
Notched Lapel | |
Serged | |
Laboratories, Remediation, Decontamination, Pesticides | |
95.25 cm |