missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isotemp™ General Purpose Laboratory Freezers
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Freezers are designed for everyday laboratory needs.
Supplier: Fisherbrand™ FBG49FSSA
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Green, natural refrigerants
- Adaptive defrost
- Digital Controller
- Rear access port
- Remote alarm contacts
- Keyed door lock
- Low heat output
- Audible and visual alarms (Temperature Hi-Lo, Door Ajar, Power Failure)
- Cord length: 10 ft.
- Sound pressure level: 51.2 dBA
- Vapor Compression System
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
General Purpose Laboratory Grade Freezer | |
1386 L | |
Solid | |
Stainless Steel | |
Stainless Steel | |
100 lb. | |
ENERGYSTAR, Prop 65, cULus | |
Forced Tube and Forced Convection Cooled | |
1 Rear Access Port | |
Adaptive Defrost | |
52.7 x 32 x 81 in. (1339 x 813 x 2057 mm) | |
Key Lock | |
11.0 A | |
115 V | |
60 Hz | |
Energy Efficient | |
R290 | |
271 kg |
49 cu. ft. | |
-19°C to -27°C | |
2 | |
Stainless Steel | |
8 | |
45 kg | |
Yes | |
Digital Microprocesor | |
3 years parts and labor, plus 3-year compressor warranty | |
FBG49FSSA | |
49 x 28.5 x 61.75 in. (1245 x 724 x 1569 mm) | |
Yes, Keyed | |
NEMA 5-15 P, three prong | |
115 V, 60 Hz | |
1 phase | |
Single Speed Hermetic | |
597 lb. |