missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isotemp™ General Purpose Laboratory Freezers
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Freezers are designed for everyday laboratory needs.
Supplier: Fisherbrand™ FBG25FSSA
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Green, natural refrigerants
- Adaptive defrost
- Digital Controller
- Rear access port
- Remote alarm contacts
- Keyed door lock
- Low heat output
Specifications
25 cu. ft. | |
-12°C to -30°C | |
Solid | |
Stainless Steel | |
Stainless Steel | |
100 lb. | |
ENERGYSTAR, Prop 65, cULus | |
Digital Microprocesor | |
2 years parts and labor, plus 3-year compressor warranty | |
27.75 x 32 x 81 in. (705 x 813 x 2057 mm) | |
Key Lock | |
NEMA 5-15 P | |
115V 60Hz | |
Energy Efficient | |
392 lb. |
700 L | |
−23°C | |
1 | |
Stainless Steel | |
4 | |
45 kg | |
Yes | |
1 Rear Access Port | |
Adaptive Defrost | |
24 x 28.5 x 61.75 in. (610 x 724 x 1569 mm) | |
8.8 A | |
115 V | |
60 Hz | |
R290 | |
178 kg |