missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl-β-D-Thiogalactopyranoside, >99%, MP Biomedicals
Molecular Biology Reagent. Cell culture media component for use in molecular genetics.β-Galactoside inducer
Supplier: MP Biomedicals Inc 0219402983
Description
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Isopropyl-β-D-Thiogalactopyranoside | |
>99% | |
MFCD00063273 | |
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
238.30 | |
CHEBI:61448 |
367-93-1 | |
C9H18O5S | |
250 mg | |
BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol | |
656894 | |
238.3 |
Chemical Identifiers
367-93-1 | |
238.30 | |
BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
656894 | |
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
C9H18O5S | |
MFCD00063273 | |
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactosideShow More | |
CHEBI:61448 |
Safety and Handling
Recommended Storage : 0°C