Learn More
Iodomethane, 99%, stabilized
CAS: 74-88-4 | CH3I | 141.94 g/mol
Supplier: Thermo Scientific Chemicals 122370050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent- Environmental benefits include:
- Renewable Energy
Specifications
Iodomethane | |
-66.0°C | |
42.5°C | |
98.5% min. (GC) | |
CH3I | |
CH3I | |
5 g | |
01,682; 02,274; 03,202; 04,341; 05,447; 06,384; 08,342; 09,308; 15,118 | |
15, 6159 | |
Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide | |
CI | |
141.94 | |
6328 | |
CHEBI:39282 | |
99% |
74-88-4,7440-50-8 | |
2.2800g/mL | |
Authentic | |
Glass bottle | |
1.5290 to 1.5310 | |
MFCD00001073 | |
01, 69 | |
2.28 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodek | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
iodomethane | |
Copper | |
0.5 mPa.s (20°C) | |
141.94 | |
Liquid |
Chemical Identifiers
74-88-4 | |
141.94 | |
INQOMBQAUSQDDS-UHFFFAOYSA-N | |
6328 | |
iodomethane |
CH3I | |
MFCD00001073 | |
methyl iodide, monoiodomethane, methane, iodo, methyljodid, jod-methan, methyliodide, methyljodide, iodometano, joodmethaan, metylu jodekShow More | |
CHEBI:39282 | |
CI |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of s
GHS Signal Word: Danger
EINECSNumber : 200-819-5
RUO – Research Use Only