missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hypromellose, Substitution Type 2910, 4000 mPa.s, USP, 7-12%, Spectrumâ„¢ Chemical
H1335, 9004-65-3
$371.17 - $2873.83
Chemical Identifiers
CAS | 9004-65-3 |
---|---|
Molecular Formula | C56H108O30 |
Molecular Weight (g/mol) | 1261.45 |
InChI Key | PUSNGFYSTWMJSK-UHFFFAOYNA-N |
IUPAC Name | 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxaneShow More |
SMILES | COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O |
Description
Spectrumâ„¢ Chemical Hypromellose, Substitution Type 2910, 4000 mPa.s, USP is an artificial tear used to relieve dry eye conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
9004-65-3 | |
1261.45 | |
1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxaneShow More |
C56H108O30 | |
PUSNGFYSTWMJSK-UHFFFAOYNA-N | |
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O |
Specifications
9004-65-3 | |
C56H108O30 | |
PUSNGFYSTWMJSK-UHFFFAOYNA-N | |
1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane | |
7 to 12% | |
1.5% | |
Poly Bottle |
100% | |
125 g | |
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O | |
1261.45 | |
USP | |
5% |