missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hypromellose, Substitution Type 2208, 3,550 mPa.s, USP, 4-12%, Spectrum™ Chemical

HY124, 9004-65-3
$371.13 - $26329.63
Chemical Identifiers
CAS | 9004-65-3 |
---|---|
Molecular Formula | C56H108O30 |
Molecular Weight (g/mol) | 1261.45 |
InChI Key | PUSNGFYSTWMJSK-UHFFFAOYNA-N |
IUPAC Name | 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane |
SMILES | COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18603386
|
Spectrum Chemical Mfg Cor
HY124125GM |
125 g | Poly Bottle |
Each for $371.13
|
|
|||||
18603387
|
Spectrum Chemical Mfg Cor
HY124500GM |
500 g | Poly Pail |
Each for $948.70
|
|
|||||
18603388
|
Spectrum Chemical Mfg Cor
HY12425KG |
2.5 kg | Poly Pail |
Each for $2,873.50
|
|
|||||
18603389
|
Spectrum Chemical Mfg Cor
HY12412KG |
12 kg | Fiber Drum |
Each for $8,991.93
|
|
|||||
18603390
|
Spectrum Chemical Mfg Cor
HY12450KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
Spectrum™ Chemical Hypromellose, Substitution Type 2208, 3,550 mPa.s, USP is an artificial tear used to relieve dry eye conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
9004-65-3 | |
1261.45 | |
1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane |
C56H108O30 | |
PUSNGFYSTWMJSK-UHFFFAOYNA-N | |
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O |
Specifications
9004-65-3 | |
C56H108O30 | |
PUSNGFYSTWMJSK-UHFFFAOYNA-N | |
1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane | |
4 to 12% | |
1.5% | |
Poly Bottle |
100% | |
125 g | |
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O | |
1261.45 | |
USP | |
5% |