missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hypromellose, Substitution Type 2208, 100 mPa.s, USP, 4-12%, Spectrum™ Chemical

HY123, 9004-65-3
$368.87 - $22902.47
Chemical Identifiers
CAS | 9004-65-3 |
---|---|
Molecular Formula | C56H108O30 |
Molecular Weight (g/mol) | 1261.45 |
InChI Key | PUSNGFYSTWMJSK-UHFFFAOYNA-N |
IUPAC Name | 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane |
SMILES | COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18603374
|
Spectrum Chemical Mfg Cor
HY123125GM |
125 g | Poly Bottle |
Each for $368.87
|
|
|||||
18603375
|
Spectrum Chemical Mfg Cor
HY123500GM |
500 g | Poly Pail |
Each for $948.70
|
|
|||||
18603376
|
Spectrum Chemical Mfg Cor
HY12325KG |
2.5 kg | Poly Pail |
Each for $2,873.50
|
|
|||||
18603377
|
Spectrum Chemical Mfg Cor
HY12312KG |
12 kg | Fiber Drum |
Each for $8,991.93
|
|
|||||
18603378
|
Spectrum Chemical Mfg Cor
HY12345KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
Spectrum™ Chemical Hypromellose, Substitution Type 2208, 100 mPa.s, USP is a semisynthetic, viscoelastic polymer that is inert and used as an ophthalmic lubricant, a controlled-delivery component and an excipient in oral medicaments. It is also used as a food additive and acts as an emulsifier, suspending agent and thickening agent. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
9004-65-3 | |
1261.45 | |
1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane |
C56H108O30 | |
PUSNGFYSTWMJSK-UHFFFAOYNA-N | |
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O |
Specifications
9004-65-3 | |
C56H108O30 | |
PUSNGFYSTWMJSK-UHFFFAOYNA-N | |
1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane | |
4 to 12% | |
1.5% | |
Poly Bottle |
100% | |
125 g | |
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O | |
1261.45 | |
USP | |
5% |