missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.1 N standard solution, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 124200100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Volumetric standard solutionSpecifications
Hydrochloric Acid | |
7647-01-0,7732-18-5 | |
1.0000g/mL | |
1 | |
ClH | |
MFCD00011324 | |
15, 4818 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene | |
[H+].[Cl-] | |
36.46 | |
CHEBI:17883 |
0.1 N Standard Solution | |
0.0998 to 0.1002N (20°C) | |
Plastic Drum | |
10L | |
HCl | |
04,450; 05,533; 06,283 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46 |
RUO â Research Use Only