Learn More
Honeywell™ Howard Leight™ Sync™ Digital AM/FM Radio
High quality digital AM/FM radio reception
Supplier:  Honeywell™ 1030331
Catalog No. 19-053-134
Only null left
Description
Description
  • Digital tuning with LCD display automatically searches for radio stations
  • 10 pre-set stations and volume memory
  • Crisp digital radio reception and high-fidelity stereo sound quality
  • Lightweight slim earcup design
  • Features patented Air Flow Control technology™ for optimal attenuation
  • Snap-in ear cushions for easy maintenance and replacement
  • Padded diamond-patterned headband with reinforced fork slides
  • Maximum volume output of FM radio at 82 dBA (based on input of 120 mV)
  • Uses 2 AA batteries with battery lifetime of 111 hours
  • Includes 3.5mm aux input cable
  • SNR 29 (H 29, M 27, L 22)
Specifications
Specifications
Digital AM and FM Radio
Product Suggestions
Product Suggestions