Learn More
Honeywell™ Howard Leight™ SmartFit™ Detectable Ear Plugs
GREENER_CHOICE
SmartFit™ detectable ear plugs for special application requirements. Conforming Material Technology™ uses body heat to adapt to the shape of the ear canal for a comfortable, personalized fit.
$153.66 - $1362.58
Specifications
ANSI S3.1974 Yes
Cord Color Blue
CSA Z94.2 Class AL
Size Regular
For Use With (Application) Food Processing, Manufacturing, Pulp and Paper, Pharmaceutical, Tobacco Processing Industries and Other Various Industries That Require Detectable Products to Prevent Contamination
View More Specs
Products 3
Catalog Number Mfr. No. Plug Color Cord Material Price Quantity  
Catalog Number Mfr. No. Plug Color Cord Material Price Quantity  
29-635-4670
View Documents
Honeywell™
SMF30BU
Blue Nylon
Pack of 100 Pair for $153.66
Case of 5 Pack for $725.00
Only null left
29-635-4669
View Documents
Honeywell™
SDT30
Blue TPE, steel
Pack of 100 Pair for $187.00
Case of 5 Pack for $882.30
Only null left
29-635-4673
View Documents
Honeywell™
SMF30
Orange Nylon
Case of 1000 Each for $1,362.58
Only null left
Description
Description
  • Patented Conforming Material Technology uses body heat to adapt to the individual shape of each wearer’s ear canal
  • Flat-fin seal with triple-fin design creates a better, more comfortable seal
  • Blue color visibility and metal ring on stem provide detection capabilities in required industries
Specifications
Specifications
Yes
Class AL
Food Processing, Manufacturing, Pulp and Paper, Pharmaceutical, Tobacco Processing Industries and Other Various Industries That Require Detectable Products to Prevent Contamination
100 Pairs/Pk.
Corded
Blue
Regular
Yes
25 dB
Flanged
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'