missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Fendall Saline Cartridges for Honeywell Pureflow 1000 Portable Emergency Eyewash Stations

Sperian Saline Cartridges

Supplier:  Honeywell™ 320010500000H5

Catalog No. 18-999-265B


Only null left
Explore available promotions

Includes:

Nozzle assembly, delivery hose, and solution cartridges


Description

Description

  • Each cartridge contains 3.5 gal. (13.2L) of saline solution
  • Tri-lingual (English, Spanish, French) 24-month cartridge life

Order Info

Each case contains two cartridges.

Specifications

Specifications

447.99 oz.
Fendall 1000 Portable Emergency Eyewash Station
Eyewash Cartridge
69 kg
13248.64 mL
Nozzle Assembly, Delivery Hose and Solution Cartridges
152.1 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions