Learn More
Honeywell™ NorthFlex Duro Task™ Textured Latex Coated Seamless Knit Gloves
Gloves feature a seamless knit shell and textured latex coating on the palm and fingertips for enhanced grip capabilities.
Supplier:  Honeywell™ NF147SDUP
Catalog No. 29-635-7282
Only null left
Explore available promotions
Alert:
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
  • Rough latex coating enhances grip capabilities
  • Seamless cotton/polyester construction increases user comfort and finger sensitivity
  • Ideal for general purpose applications such as assembly work, construction, and warehousing
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Product Suggestions
Product Suggestions