missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazardous Material Label: ISOPROPYL ALCOHOL

Provide workers with all of the chemical information needed for hazardous materials they may encounter. Brady™ Hazardous Material Labels are made of vinyl and display text in black on yellow.

Supplier:  Brady™ 93554

Catalog No. 19-107-676


Only null left
Explore available promotions

Description

Description

  • Provides workers with chemical information of hazardous materials
  • Made from B-928 vinyl material
  • Label Size: W x H: 4.5 x 3.75 in.
Specifications

Specifications

Acrylic, Permanent, Cold-temperature
Vinyl (B-928)
ISOPROPYL ALCOHOL
Rectangle
0.0049 in.
Black, Yellow
Hazardous Materials Label
25/Pk.
4.5 x 3.75 in. (W x H)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions