missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium wire, 1.0mm (0.04in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 010205CC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Hafnium wire | |
7440-58-6 | |
Odorless | |
Wire | |
Hf | |
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire | |
[Hf] | |
178.49 | |
CHEBI:33343 |
1.0mm (0.04 in.) dia. | |
500 cm | |
≈10.28g/m | |
(metals basis excluding Zr), Zr nominal 3% | |
MFCD00011032 | |
VBJZVLUMGGDVMO-UHFFFAOYSA-N | |
hafnium | |
23986 | |
99.95% |
Chemical Identifiers
7440-58-6 | |
178.49 | |
VBJZVLUMGGDVMO-UHFFFAOYSA-N | |
23986 | |
hafnium |
Hf | |
MFCD00011032 | |
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire | |
CHEBI:33343 | |
[Hf] |
Safety and Handling
EINECSNumber : 231-166-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only