Learn More
Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis)
Form: Wire cloth; Open area: 62.7%; Width of opening: 0.015 in. | CAS: 7440-57-5 | Au | 196.97 g/mol
Supplier: Thermo Scientific Chemicals 040930GH
Description
Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis) is used as an electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Certifications and Seals of Approval
Specifications
Golden | |
7440-57-5 | |
99.99% (Metals basis) | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
52 Mesh | |
Wire Cloth |
Gold | |
Gold colored gauze | |
Au | |
100 x 100 mm | |
Soluble in aqua regia. | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Odorless |
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Safety and Handling
EINECSNumber : 231-165-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only