missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaric dialdehyde, 25% w/v solution in water
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Supplier: Thermo Scientific Chemicals 119980100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glutaric dialdehyde | |
-5.0°C | |
3.2 to 4.2 | |
Plastic drum | |
1.373 | |
MFCD00007025 | |
01, 776 | |
1.06 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.12 | |
Solution |
111-30-8,7732-18-5 | |
1.0600g/mL | |
101°C | |
C5H8O2 | |
OHC(CH2)3CHO | |
10 L | |
01,411 | |
15, 4508 | |
Solubility in water: soluble | |
O=CCCCC=O | |
100.12 | |
CHEBI:64276 | |
16.4mmHg at 20°C |
RUO – Research Use Only