missing translation for 'onlineSavingsMsg'
Learn More

Glutaric anhydride, 95%

CAS: 108-55-4 | C5H6O3 | 114.1 g/mol

$69.70 - $590.24

Chemical Identifiers

CAS 108-55-4
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00006679
InChI Key VANNPISTIUFMLH-UHFFFAOYSA-N
Synonym glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czechShow MoreShow Less
PubChem CID 7940
IUPAC Name oxane-2,6-dione
SMILES C1CC(=O)OC(=O)C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119970050
SDS View Documents
Thermo Scientific Chemicals
119970050
5 g Glass bottle
Each for $69.70
Only null left
 
AC119971000
SDS View Documents
Thermo Scientific Chemicals
119971000
100 g Glass bottle
Each for $155.57
Only null left
 
AC119975000
SDS View Documents
Thermo Scientific Chemicals
119975000
500 g Glass bottle
Each for $590.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 203-593-6

RTECSNumber : MA3850000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only