Learn More
Glutaric acid, 99%

CAS: 110-94-1 | C5H8O4 | 132.12 g/mol
$62.04 - $445.15
Chemical Identifiers
CAS | 110-94-1 |
---|---|
Molecular Formula | C5H8O4 |
Molecular Weight (g/mol) | 132.12 |
MDL Number | MFCD00004410 |
InChI Key | JFCQEDHGNNZCLN-UHFFFAOYSA-N |
Synonym | glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-diShow More |
PubChem CID | 743 |
ChEBI | CHEBI:17859 |
IUPAC Name | pentanedioic acid |
SMILES | OC(=O)CCCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC119960250
|
Thermo Scientific Chemicals
119960250 |
25 g | Glass Bottle |
N/A
|
|
|||||
AC119961000
|
Thermo Scientific Chemicals
119961000 |
100 g | Plastic bottle |
N/A
|
|
|||||
AC119965000
|
Thermo Scientific Chemicals
119965000 |
500 g | Plastic Bottle |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-94-1 | |
132.12 | |
JFCQEDHGNNZCLN-UHFFFAOYSA-N | |
743 | |
pentanedioic acid |
C5H8O4 | |
MFCD00004410 | |
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-diShow More | |
CHEBI:17859 | |
OC(=O)CCCC(O)=O |
Specifications
110-94-1 | |
Orange | |
Authentic | |
Glass Bottle | |
HO2C(CH2)3CO2H | |
25 g | |
01,412 | |
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di | |
JFCQEDHGNNZCLN-UHFFFAOYSA-N | |
pentanedioic acid | |
743 | |
132.12 | |
Crystalline Powder |
92.0°C to 99.0°C | |
302.0°C | |
99% | |
C5H8O4 | |
MFCD00004410 | |
02, 631 | |
15, 4509 | |
Solubility in water: 430g/L (20°C). Other solubilities: freely soluble in abs. alcohol and ether, soluble in benzene and chloroform | |
OC(=O)CCCC(O)=O | |
132.12 | |
CHEBI:17859 | |
99% | |
Glutaric acid, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISO
GHS Signal Word: Warning
EINECSNumber : 203-817-2
RTECSNumber : MA3740000
TSCA : TSCA
RUO – Research Use Only