missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glucagon Like Peptide-1 (7-37)
GREENER_CHOICE

For cellular and molecular biology applications

$365.42 - $695.53

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Lyophilized Powder
Molecular Weight (g/mol) 3355.70
Product Type Glucagon Like Peptide-1 (7-37)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66232EXD
View Documents
Thermo Scientific Chemicals
J66232EXD
0.5 mg
Each for $365.42
Only null left
 
AAJ66232MCR
View Documents
Thermo Scientific Chemicals
J66232MCR
1 mg
Each for $695.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Lyophilized Powder
Glucagon Like Peptide-1 (7-37)
-30°C to -10°C
3355.70
RUO - research use only
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only