Learn More
World Precision Standard Glass Capillaries-1B120F-4 DFS Item
Supplier:  World Precision 1B120F4
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.
Catalog No. 50-821-813
Only null left
Explore available promotions