Learn More
Gallocyanine
CAS: 1562-85-2 | C15H13ClN2O5 | 336.72 g/mol
Supplier: Thermo Scientific Chemicals 154430100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gallocyanine | |
Black to Green | |
Glass bottle | |
MFCD00011926 | |
27, 438 | |
gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue p | |
CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-] | |
336.72 | |
336.72 |
1562-85-2 | |
Authentic | |
C15H13ClN2O5 | |
10 g | |
15, 4385 | |
AQSOTOUQTVJNMY-UHFFFAOYSA-N | |
7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride | |
73801 | |
Fine Crystalline Powder |
Chemical Identifiers
1562-85-2 | |
336.72 | |
AQSOTOUQTVJNMY-UHFFFAOYSA-N | |
73801 | |
CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-] |
C15H13ClN2O5 | |
MFCD00011926 | |
gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue pShow More | |
7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chlorideShow More |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
DOTInformation : DOT Class 6.1
EINECSNumber : 216-345-7
RTECSNumber : SP7692000
RUO – Research Use Only