Learn More
Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 444101000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysisSpecifications
Gallium(III) chloride | |
100g | |
Cl3Ga | |
15,4378 | |
Solubility in water: reacts | |
Cl[Ga](Cl)Cl | |
176.08 | |
176.08 | |
Trace Metal Basis |
13450-90-3 | |
99.99+% | |
GaCl3 | |
gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue | |
UPWPDUACHOATKO-UHFFFAOYSA-K | |
trichlorogallane | |
26010 | |
99.99+% | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Store in a d
GHS Signal Word: Danger
EINECSNumber : 236-610-
RUO â Research Use Only