missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fusaric acid, 99%
CAS: 536-69-6 | C10H13NO2 | 179.22 g/mol
$495.09 - $495.09
Chemical Identifiers
CAS | 536-69-6 |
---|---|
Molecular Formula | C10H13NO2 |
Molecular Weight (g/mol) | 179.22 |
MDL Number | MFCD00006298 |
InChI Key | DGMPVYSXXIOGJY-UHFFFAOYSA-N |
Synonym | fusaric acid, 5-butylpicolinic acid, fusarinic acid, 2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, picolinic acid, 5-butyl, 5-n-butylpyridine-2-carboxylic acid, unii-jwj963070n, 5-butylpyridine-3-carboxylic acid, 5-butyl-pyridine-2-carboxylic acidShow More |
PubChem CID | 3442 |
ChEBI | CHEBI:5199 |
IUPAC Name | 5-butylpyridine-2-carboxylic acid |
SMILES | CCCCC1=CN=C(C=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC198960010
|
Thermo Scientific Chemicals
198960010 |
1 g | Glass bottle |
Each for $495.09
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
536-69-6 | |
179.22 | |
DGMPVYSXXIOGJY-UHFFFAOYSA-N | |
3442 | |
5-butylpyridine-2-carboxylic acid |
C10H13NO2 | |
MFCD00006298 | |
fusaric acid, 5-butylpicolinic acid, fusarinic acid, 2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, picolinic acid, 5-butyl, 5-n-butylpyridine-2-carboxylic acid, unii-jwj963070n, 5-butylpyridine-3-carboxylic acid, 5-butyl-pyridine-2-carboxylic acidShow More | |
CHEBI:5199 | |
CCCCC1=CN=C(C=C1)C(=O)O |
Specifications
536-69-6 | |
White to Yellow | |
99% | |
C10H13NO2 | |
1 g | |
fusaric acid, 5-butylpicolinic acid, fusarinic acid, 2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, picolinic acid, 5-butyl, 5-n-butylpyridine-2-carboxylic acid, unii-jwj963070n, 5-butylpyridine-3-carboxylic acid, 5-butyl-pyridine-2-carboxylic acid | |
CCCCC1=CN=C(C=C1)C(=O)O | |
179.22 | |
CHEBI:5199 | |
99% | |
Fusaric acid |
97°C to 101°C | |
Authentic | |
Glass bottle | |
MFCD00006298 | |
15, 4343 | |
DGMPVYSXXIOGJY-UHFFFAOYSA-N | |
5-butylpyridine-2-carboxylic acid | |
3442 | |
179.22 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Warning
EINECSNumber : 208-643-
RUO – Research Use Only