Learn More
Formamide, 99.5%, for analysis
CAS: 75-12-7 | CH3NO | 45.04 g/mol
Supplier: Thermo Scientific Chemicals 205820010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis- Environmental benefits include:
- Recyclable
Specifications
Formamide | |
75-12-7 | |
210.0°C | |
175°C | |
Glass Bottle | |
1.4460 to 1.4480 | |
1.133 | |
HCONH2 | |
3.75 mPa.s (20°C) | |
02, 26 | |
15, 4265 | |
Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene | |
NC=O | |
45.04 | |
CHEBI:48431 | |
99.5% |
2.0°C to 3.0°C | |
1.1330g/mL | |
1 L | |
Authentic | |
Liquid | |
99.5% | |
CH3NO | |
MFCD00007941 | |
0.05% max. (Coulometric) | |
01,402; 02,201; 03,147 | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
formamide | |
713 | |
45.04 | |
Analytical |
Chemical Identifiers
75-12-7 | |
45.04 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
713 | |
formamide |
CH3NO | |
MFCD00007941 | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
CHEBI:48431 | |
NC=O |
Safety and Handling
GHS P Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. Suspected of causing cancer if swallowed.
GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required.
Danger
RUO – Research Use Only