Learn More
Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 440360250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentSpecifications
Fluoro-N,N,N′,N′-bis(tetramethylene)formamidinium hexafluorophosphate | |
32.82 to 35.55% C, 8.51 to 9.21% N | |
96% min. (HPLC) | |
C9H16FN2·F6P | |
btffh, bis tetramethylene fluoroformamidinium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, c9h16fn2.f6p, fluoro-dipyrrolidinocarbenium hexafluorophosphate, 1-fluoro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate t, 1e-1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n inverted exclamation marka,n inverted exclamation marka-bis tetramethylene formamidinium hexafluorophosphate | |
C1CCN(C1)C(=[N+]2CCCC2)F.F[P-](F)(F)(F)(F)F | |
316.2 | |
316.2 |
164298-25-3 | |
Authentic | |
Glass bottle | |
25g | |
MNJUGQKOHJQOCK-UHFFFAOYSA-N | |
1-[fluoro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate | |
10935980 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
RUO â Research Use Only