Learn More
Fluoride Standard, 2 ppm F- (Premixed with TISAB/Color Coded Red), Ricca Chemical
Supplier:  Ricca Chemical Company 31631
Catalog No. 31631
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Specifications
Specifications
Fluoride Standard
4 L
93.0,1.94,1.63,1.58,0.11,0.01
97.0,2.02,1.69,1.64,0.11,0.01
Natural Poly Bottle
7681-49-4,1310-73-2,64-19-7,7647-14-5,13291-61-7,25956-17-6, 7732-18-5
2ppm F-
1 mL = 0.002 mg F-
Product Suggestions
Product Suggestions