Learn More
Greiner Bio-One Flat Bottom Polystyrene Multipurpose Container with Metal Screw Cap
GREENER_CHOICE
Ideal for optical measurements due to its high clarity.
$334.60 - $822.70
Specifications
Product Type Multi-purpose Container
Material Polystyrene
Closure Material Metal
Closure Type Screw Cap
For Use With (Application) For academic and medical purposes
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Sterility Label Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Sterility Label Price Quantity  
07000487
View Documents
Greiner Bio-One
224170
100 mL Aseptic No Label
Case of 160 Each for $822.70
Only null left
07000491
View Documents
Greiner Bio-One
225180
150 mL Aseptic Labeled
Case of 120 Each for $799.27
Only null left
07000502
View Documents
Greiner Bio-One
229170
250 mL Aseptic No Label
Case of 48 Each for $334.60
Only null left
Description
Description
  • Metal screw cap
  • With or without plain label
  • Manufactured aseptically in a Class II Clean Room environment
  • Can be used universally for academic and medical purposes
Specifications
Specifications
Multi-purpose Container
Metal
For academic and medical purposes
Polystyrene
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'