Learn More
Chemglass Life Sciences 50mL Single Neck Evaporating Flask, 24/40 Outer Joint, Recovery Flask DFS Item
Supplier:  Chemglass Inc CG151201
50mL Single Neck Evaporating Flask, 24/40 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.
Catalog No. CG151201
Only null left
Explore available promotions
Description
Description
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
    • Ozone-Safe or Safe for the Ozone Layer
Product Suggestions
Product Suggestions